Phenethyl bromide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(2-Bromoethyl)benzene, 2-Phenylethyl bromide
10694-0025
10694-1000
10694-5000
TS10694-0025EA
528
USD
TS10694-0025
TS10694-1000
TS10694-5000
Phenethyl bromide 98%
Phenethyl bromide
Formula:
BrCH₂CH₂C₆H₅ MW: 185.06 g/mol Boiling Pt: 220…221 °C (760 mmHg) Melting Pt: –56 °C Density: 1.35 g/cm³ Flash Pt: 89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000240 CAS Number: 103-63-9 EINECS: 203-130-8 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5553 to 1.5573 (20°C, 589 nm) |
Water | ≤0.05 % |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...