1-Bromo-4-chlorobutane 99%

Supplier: Thermo Scientific Chemicals
Warning

10684-0250 10684-1000
TS10684-0250EA 43 USD
TS10684-0250 TS10684-1000
1-Bromo-4-chlorobutane 99%
1-Bromo-4-chlorobutane
Formula: BrH₂C(CH₂)₂CH₂Cl
MW: 171.46 g/mol
Boiling Pt: 80…82 °C (30 mmHg)
Density: 1.48 g/cm³
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00001010
CAS Number: 6940-78-9
EINECS: 230-089-3
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4865 to 1.4885 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR