2-Bromobutane 99+%

Supplier: Thermo Scientific Chemicals
Danger

10678-0025 10678-1000 10678-5000
TS10678-0025EA 338 USD
TS10678-0025 TS10678-1000 TS10678-5000
2-Bromobutane 99+%
2-Bromobutane
Formula: H₃CBrCHCH₂CH₃
MW: 137.02 g/mol
Boiling Pt: 91 °C (760 mmHg)
Melting Pt: –112 °C
Density: 1.25 g/cm³
Flash Pt: 21 °C
Storage Temperature: Ambient
MDL Number: MFCD00000156
CAS Number: 78-76-2
EINECS: 201-140-7
UN: 2339
ADR: 3 II
Merck Index: 13,01553

Order Now

Specification Test Results

Appearance Clear colorless to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.4359 to 1.4379 (20°C, 589 nm)
GC ≥99.0 %
Titration with HClO4 ≥98.5 %
Sensitivity (Test for Ni) Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR