Bromoacetyl bromide 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
10660-1000
10660-5000
TS10660-1000EA
149
USD
TS10660-1000
TS10660-5000
Bromoacetyl bromide 98%
Bromoacetyl bromide
Formula:
BrCH₂COOBr Boiling Pt: 148 °C (1013 hPa) Density: 2.31 g/cm³ Flash Pt: >105 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000115 CAS Number: 598-21-0 EINECS: 209-923-5 UN: 2513 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to yellow Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.5450 to 1.5490 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...