4,4'-Anisoin 95%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Hydroxy-1,2-bis(4-methoxyphenyl)ethanone, Anisoin

10484-0250 10484-1000
TS10484-0250EA 53 USD
TS10484-0250 TS10484-1000
4,4'-Anisoin 95%
4,4'-Anisoin
Formula: C₁₆H₁₆O₄
MW: 272.30 g/mol
Melting Pt: 109…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00008411
CAS Number: 119-52-8
EINECS: 204-330-8

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
Melting point 108 to 115 °C
HPLC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR