Allyl isothiocyanate 94% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with α-tocopherol 0.01%
10295-0050
10295-1000
10295-5000
TS10295-0050EA
28
USD
TS10295-0050
TS10295-1000
TS10295-5000
Allyl isothiocyanate 94% stabilized
Allyl isothiocyanate
Formula:
C₄H₅NS MW: 99.16 g/mol Boiling Pt: 151…153 °C (760 mmHg) Melting Pt: –80 °C Density: 1.0175 g/cm³ Flash Pt: 46 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004822 CAS Number: 57-06-7 EINECS: 200-309-2 UN: 1545 ADR: 6.1,II Merck Index: 13,00292 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5300 to 1.5330 (20°C, 589 nm) |
GC | ≥92 % |
Stabilizer | 0.01 % alpha-Tocopherol |
Relative density | (20/4°C) 1.017 to 1.022 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...