Uridine-5′-monophosphate disodium hydrate 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
5'-Uridylic acid disodium salt xhydrate, Uridylic acid disodium salt hydrate, 5'-Uridylic acid disodium salt hydrate, U 5′-P hydrate, UMP hydrate, disodium [(2R,3S,4S,5R)-5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate hydrate, Uridine 5‘-monophosphate sel disodique hydrate, 5′-UMP-Na2 hydrate
22629-0010
22629-0050
22629-0250
TS22629-0010EA
35
USD
TS22629-0010
TS22629-0050
TS22629-0250
Uridine-5′-monophosphate disodium hydrate 98%
Uridine-5′-monophosphate disodium hydrate
Formula:
C₉H₁₁N₂Na₂O₉P·xH₂O MW: 386.160 g/mol Melting Pt: 208…210 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149422 CAS Number: 681435-27-8 |
Specification Test Results
Appearance | White Powder |
Infrared spectrum | Conforms |
Water | ≤26 % (K.F.) |
HPLC | ≥97.5 % |
UV spectrophotometry | molar absorptivity 10000 +/- 2%, on dry basis |
UV transmittance | ≥95 % (c=1, 430 nm, in H2O) 1 cm cell |
Ratio of Absorbances | 0.38 to 0.44 (A280/A260) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...