Methyl-β-D-thiogalactoside 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: TMG, Thiomethyl β-D-galactopyranoside

45945-0010 45945-2500
TS45945-0010EA 281 USD
TS45945-0010 TS45945-2500
Methyl-β-D-thiogalactoside 98%
Methyl-beta-D-thiogalactoside
Formula: C₇H₁₄O₅S
MW: 210.25 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00038073
CAS Number: 155-30-6

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 176 to 180 °C
Water ≤1 %
HPLC ≥97.5 %
Specific optical rotation +10° to +12° (20 °C, 589 nm) (5 % in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR