Perfluoro-compound FC-3283™

Supplier: Thermo Scientific Chemicals

Synonyms: Fluorinert® FC-3283

45858-1000 45858-2500
TS45858-1000EA 138 USD
TS45858-1000 TS45858-2500
Perfluoro-compound FC-3283™
Perfluoro-compound FC-3283™
MW: 521.00 g/mol
Boiling Pt: 123…133 °C (760 mmHg)
Density: 1.8 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD01632188
CAS Number: 316806-89-0

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Boiling point 123 to 133 °C
Density 1.780 to 1.860 kg/l
Viscosity 0.70 to 0.90 cSt (at 25 °C)
Dielectric strength ≥35 kV
Distillation range ≥95 % (between 123-133 °C)
Pour point < -40 °C
Residue ≤10 ug/ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR