3-Acetylpyridine adenine dinucleotide 90%

Supplier: Thermo Scientific Chemicals

Synonyms: APAD, 3-Acetyl-1-((2R,3R,4S,5R)-5-((((((((2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)oxidophosphoryl)oxy)methyl)-3,4-dihydroxytetrahydrofuran-2-yl)pyridin-1-ium

45768-0010 45768-2500
TS45768-0010EA 1154 USD
TS45768-0010 TS45768-2500
3-Acetylpyridine adenine dinucleotide 90%
3-Acetylpyridine adenine dinucleotide
Formula: C₂₂H₂₈N₆O₁₄P₂
MW: 662.44 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00078882
CAS Number: 86-08-8

Order Now

Specification Test Results

Appearance White to off-white to light yellow Powder
Infrared spectrum Conforms
Water ≤8 %
HPLC ≥89.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR