4,4,4-Trifluorobutylamine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Butanamine,4,4,4-trifluoro-, 4,4,4-trifluorobutan-1-amine, 1-Amino-4,4,4-trifluorobutane

43259-0010 43259-0050
TS43259-0010EA 157 USD
TS43259-0010 TS43259-0050
4,4,4-Trifluorobutylamine 97%
4,4,4-Trifluorobutylamine
Formula: C₄H₈F₃N
MW: 127.11 g/mol
Boiling Pt: 90…94 °C (760 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD06213030
CAS Number: 819-46-5
UN: 2734
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥96.0 %
Titration RCOCl ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR