2-(Tetrazol-5-yl)phenylboronic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(5-Tetrazolyl)phenylboronic acid, (2-(1H-Tetrazol-5-yl)phenyl)boronic acid, Boronic acid,B-[2-(2H-tetrazol-5-yl)phenyl]-, 2-(1H-Tetrazol-5-yl)phenylboronic acid

44555-0010 44555-0050
TS44555-0010EA 119 USD
TS44555-0010 TS44555-0050
2-(Tetrazol-5-yl)phenylboronic acid 95%
2-(Tetrazol-5-yl)phenylboronic acid
Formula: C₇H₇BN₄O₂
Melting Pt: 148…152 °C
Storage Temperature: Ambient
MDL Number: MFCD06796325
CAS Number: 155884-01-8

Order Now

Specification Test Results

Appearance White to off-white Solid
Infrared spectrum Conforms
Additional info May contain varying amounts of anhydrides
HPLC ≥94.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR