Potassium (cyanomethyl)trifluoroborate 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Potassium (cyanomethyl)(trifluoro)borate(1-), potassium (cyanomethyl)trifluoroboranuide

44486-0010 44486-0050
TS44486-0010EA 224 USD
TS44486-0010 TS44486-0050
Potassium (cyanomethyl)trifluoroborate 95%
Potassium (cyanomethyl)trifluoroborate
Formula: C₂H₂BF₃KN
MW: 146.949 g/mol
Melting Pt: 151…156 °C
Storage Temperature: Ambient
MDL Number: MFCD11052818
CAS Number: 888711-58-8
UN: 3439
ADR: 6.1 III

Order Now

Specification Test Results

Appearance White to off-white Solid
Infrared spectrum Conforms
Melting point 144 to 156 °C
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR