3-(N-Boc-aminomethyl)-5-bromopyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Carbamic acid,N-[(5-bromo-3-pyridinyl)methyl]-, 1,1-dimethylethyl ester, 3-Bromo-5-{[(tert-butoxycarbonyl)amino]methyl}pyridine, tert-Butyl ((5-bromopyridin-3-yl)methyl)carbamate, tert-Butyl (5-bromopyridin-3-yl)methylcarbamate
44406-0010
44406-0050
TS44406-0010EA
262
USD
TS44406-0010
TS44406-0050
3-(N-Boc-aminomethyl)-5-bromopyridine 97%
3-(N-Boc-aminomethyl)-5-bromopyridine
Formula:
C₁₁H₁₅BrN₂O₂ MW: 287.157 g/mol Melting Pt: 84.5…86 °C Storage Temperature: Ambient |
MDL Number:
MFCD10700149 CAS Number: 943722-24-5 |
Specification Test Results
Appearance | White to yellow Powder |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...