2-Chloropyrimidine-5-sulphonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
5-Pyrimidinesulphonyl chloride, 2-chloro-
42963-0010
42963-0050
TS42963-0010EA
329
USD
TS42963-0010
TS42963-0050
2-Chloropyrimidine-5-sulphonyl chloride 97%
2-Chloropyrimidine-5-sulfonyl chloride
Formula:
C₄H₂Cl₂N₂O₂S Melting Pt: 64…67 °C Storage Temperature: Freezer |
MDL Number:
MFCD06657208 CAS Number: 98026-88-1 UN: 3261 ADR: 8,III |
Specification Test Results
Appearance | White to yellow to light orange Crystalline solid |
Infrared spectrum | Conforms |
Melting point | 64 to 68 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...