3-Amino-2,2,5,5-tetramethyl-1-pyrrolidinyloxy 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Pyrrolidinyloxy, 3-amino-2,2,5,5-tetramethyl-, (3-Amino-2,2,5,5-tetramethylpyrrolidin-1-yl)oxidanyl, (3-Amino-2,2,5,5-tetramethyl-1-pyrrolidinyl)oxidanyl

40110-0020 40110-5000
TS40110-0020EA 773 USD
TS40110-0020 TS40110-5000
3-Amino-2,2,5,5-tetramethyl-1-pyrrolidinyloxy 99%
3-Amino-2,2,5,5-tetramethyl-1-pyrrolidinyloxy
Formula: C₈H₁₇N₂O
MW: 157.24 g/mol
Boiling Pt: 75…80 °C (1 mmHg)
Melting Pt: 45 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00046088
CAS Number: 34272-83-8
UN: 3263
ADR: 8,III

Order Now

Specification Test Results

Appearance Orange Hygroscopic crystals
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR