3-Formylsalicylic acid hydrate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
37707-0010
37707-0050
37707-0250
TS37707-0010EA
80
USD
TS37707-0010
TS37707-0050
TS37707-0250
3-Formylsalicylic acid hydrate 97%
3-Formylsalicylic acid hydrate
Formula:
C₈H₈O₅ Melting Pt: 249…252 °C Storage Temperature: Ambient |
MDL Number:
MFCD03804454 CAS Number: 1266615-78-4 |
Specification Test Results
Appearance | Yellow to beige to light brown Crystalline powder or needles |
Infrared spectrum | Conforms |
Loss on drying | ≤11 % (1 g, 105°C, 3 hrs) |
HPLC | ≥96.0 % |
Titration with NaOH | ≥96.0 % (On dry substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...