2-(2-Propynyloxy)tetrahydro-2H-pyran 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tetrahydro-2-(2-propynyloxy)-2H-pyran, 2-Propargyloxane, 2-(Prop-2-yn-1-yloxy)tetrahydro-2H-pyran, Propargyl 2-tetrahydropyranyl ether, 2-(2-Propynyloxy)tetrahydropyran

29506-0050 29506-0250 29506-1000
TS29506-0050EA 43 USD
TS29506-0050 TS29506-0250 TS29506-1000
2-(2-Propynyloxy)tetrahydro-2H-pyran 98%
2-(2-Propynyloxy)tetrahydro-2H-pyran
Formula: C₈H₁₂O₂
Boiling Pt: 63…65 °C (9 mmHg)
Density: 0.997 g/cm³
Flash Pt: 59 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006604
CAS Number: 6089-04-9
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4560 to 1.4580 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR