Propargyl acetate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Propargylacetate

38082-0050 38082-0250
TS38082-0050EA 44 USD
TS38082-0050 TS38082-0250
Propargyl acetate 97%
Propargyl acetate
Formula: C₅H₆O₂
Boiling Pt: 27…28 °C (8 mmHg)
Density: 0.989 g/cm³
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00041601
CAS Number: 627-09-8
UN: 1992
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4165 to 1.4195 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR