Carnauba wax, yellow, refined No. 1

Supplier: Thermo Scientific Chemicals

22238-0010 22238-0050 22238-2500
TS22238-0010EA 109 USD
TS22238-0010 TS22238-0050 TS22238-2500
Carnauba wax, yellow, refined No. 1
Carnauba wax
Melting Pt: 81…86 °C
Flash Pt: 282 °C
Storage Temperature: Ambient
MDL Number: MFCD00130724
CAS Number: 8015-86-9

Order Now

Specification Test Results

Appearance Yellow Flakes
Melting point 80 to 88°C
Residue after ignition ≤0.25 %
Acid value 2 to 7 mg KOH/g
Saponification value 78 to 95 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR