(+/-)-Usnic acid

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Usnic acid, 2,6-Diacetyl-7,9-dihydroxy-8,9b-dimethyldibenzo[b,d]furan-1,3(2H,9bH)-dione

46285-0010 46285-0050 46285-0250
TS46285-0010EA 34 USD
TS46285-0010 TS46285-0050 TS46285-0250
(+/-)-Usnic acid
(+/-)-Usnic acid
Formula: C₁₈H₁₆O₇
MW: 344.3155 g/mol
Melting Pt: 201…203 °C
Storage Temperature: Ambient
MDL Number: MFCD00065294
CAS Number: 125-46-2
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Yellow to light green or gold Powder
Infrared spectrum Conforms
Melting point 193 to 203 °C
Loss on drying ≤2 %
Heavy metals ≤10 ppm
HPLC ≥97.5 %
Residue after ignition ≤2 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR