4-Iodo-1H-pyrazol-5-amine 97%

Supplier: Thermo Scientific Chemicals
Warning

44452-0050 44452-0010
TS44452-0050EA 403 USD
TS44452-0050 TS44452-0010
4-Iodo-1H-pyrazol-5-amine 97%
4-Iodo-1H-pyrazol-5-amine
Formula: C₃H₄IN₃
Melting Pt: 143…146 °C
Storage Temperature: Ambient
MDL Number: MFCD22422021
CAS Number: 81542-51-0

Order Now

Specification Test Results

Appearance Off-white to brown Solid
Infrared spectrum Conforms
Melting point 140 to 146 °C
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR