1-Tetrahydro-2H-pyran-4-yl-1H-pyrazol-4-amine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Tetrahydropyran-4-yl-1H-pyrazol-4-ylamine, 1-(Tetrahydro-2H-pyran-4-yl)-1H-pyrazol-4-amine
45357-0010
45357-0050
TS45357-0010EA
205
USD
TS45357-0010
TS45357-0050
1-Tetrahydro-2H-pyran-4-yl-1H-pyrazol-4-amine 97%
1-Tetrahydro-2H-pyran-4-yl-1H-pyrazol-4-amine
Formula:
C₈H₁₃N₃O MW: 167.21 g/mol Melting Pt: 105…107 °C Storage Temperature: Ambient |
MDL Number:
MFCD11505007 CAS Number: 1190380-49-4 |
Specification Test Results
Appearance | White to light yellow to pink Powder |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...