5-(Chloromethyl)-1,3-dimethyl-1H-pyrazole 97%

Supplier: Thermo Scientific Chemicals
Danger

45156-0010 45156-0100
TS45156-0010EA 141 USD
TS45156-0010 TS45156-0100
5-(Chloromethyl)-1,3-dimethyl-1H-pyrazole 97%
5-(Chloromethyl)-1,3-dimethyl-1H-pyrazole
Formula: C₆H₉ClN₂
MW: 144.61 g/mol
Melting Pt: 65…66 °C
Storage Temperature: Ambient
MDL Number: MFCD07368502
CAS Number: 852227-86-2
UN: 3261
ADR: 8 III

Order Now

Specification Test Results

Appearance White to yellow Low melting solid
Infrared spectrum Conforms
Melting point 63 to 69 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR