5-Cyclopropyl-1,2,4-oxadiazol-3-amine 97%

Supplier: Thermo Scientific Chemicals
Warning

45003-0010 450030050
TS45003-0010EA 248 USD
TS45003-0010 TS450030050
5-Cyclopropyl-1,2,4-oxadiazol-3-amine 97%
5-Cyclopropyl-1,2,4-oxadiazol-3-amine
Formula: C₅H₇N₃O
Storage Temperature: Ambient
MDL Number: MFCD11847813
CAS Number: 868696-42-8

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR