2-Amino-3,3,3-trifluoropropionic acid ethylester hydrochloride 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
44664-0010
44664-0050
TS44664-0010EA
156
USD
TS44664-0010
TS44664-0050
2-Amino-3,3,3-trifluoropropionic acid ethylester hydrochloride 95%
2-Amino-3,3,3-trifluoropropionic acid ethylester hydrochloride
Formula:
C₅H₉ClF₃NO₂ Melting Pt: 218…220 °C Storage Temperature: Ambient |
MDL Number:
MFCD14525479 CAS Number: 193140-71-5 |
Specification Test Results
Appearance | White to yellow Powder or solid |
Infrared spectrum | Conforms |
Titration Argentometric | ≥94.0 to ≤106.0 % |
Water | ≤2.5 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...