3,5-Difluoro-4-pyridinamine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
45253-0010
45253-0050
TS45253-0010EA
203
USD
TS45253-0010
TS45253-0050
3,5-Difluoro-4-pyridinamine 97%
3,5-Difluoro-4-pyridinamine
Formula:
C₅H₄F₂N₂ Melting Pt: 102…105 °C Storage Temperature: Ambient |
MDL Number:
MFCD01862101 CAS Number: 159783-22-9 |
Specification Test Results
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Melting point | 100 to 105 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...