5-Bromo-1H-pyrrolo[2,3-b]pyridine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
5-Bromo-7-azaindole
43743-0010
43743-0050
TS43743-0010EA
55
USD
TS43743-0010
TS43743-0050
5-Bromo-1H-pyrrolo[2,3-b]pyridine 97%
5-Bromo-1H-pyrrolo[2,3-b]pyridine
Formula:
C₇H₅BrN₂ Melting Pt: 179.7…180.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD06659677 CAS Number: 183208-35-7 |
Specification Test Results
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 177 to 182 °C |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...