4-Bromoisoindoline hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

43587-0010 43587-0050
TS43587-0010EA 116 USD
TS43587-0010 TS43587-0050
4-Bromoisoindoline hydrochloride 97%
4-Bromoisoindoline hydrochloride
Formula: C₈H₉BrClN
Melting Pt: 269…274 °C
Storage Temperature: Ambient
MDL Number: MFCD10700134
CAS Number: 923590-95-8

Order Now

Specification Test Results

Appearance Off-white to yellow to pink to brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Elemental Analysis 5.73 to 6.21 % N

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR