Methyl p-tolyl sulphoxide 98%

Supplier: Thermo Scientific Chemicals

42815-0010 42815-0050
TS42815-0010EA 72 USD
TS42815-0010 TS42815-0050
Methyl p-tolyl sulphoxide 98%
Methyl p-tolyl sulfoxide
Formula: C₈H₁₀OS
Boiling Pt: 113…114 °C (2.2 mmHg)
Melting Pt: 41…46 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00075175
CAS Number: 934-72-5

Order Now

Specification Test Results

Appearance White to yellow Low melting powder or crystals
Infrared spectrum Conforms
Melting point 41 to 46 °C
GC ≥97.5 %
NMR consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR