Phenyl-p-benzoquinone 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Phenyl-1,4-benzoquinone, [1,1'-Biphenyl]-2,5-dione
41738-0250
41738-1000
TS41738-0250EA
121
USD
TS41738-0250
TS41738-1000
Phenyl-p-benzoquinone 95%
Phenyl-p-benzoquinone
Formula:
C₁₂H₈O₂ MW: 184.194 g/mol Melting Pt: 111…115 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001599 CAS Number: 363-03-1 |
Specification Test Results
Appearance | Yellow-brown Powder |
Infrared spectrum | Conforms |
Melting point | 111 to 115 °C |
HPLC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...