2,2'-Azobis(2-methylpropionamidine)dihydrochloride (AAPH) 98%

Supplier: Thermo Scientific Chemicals
Danger

40156-2500 40156-0250 40156-0500
TS40156-2500EA 219 USD
TS40156-2500 TS40156-0250 TS40156-0500
2,2'-Azobis(2-methylpropionamidine)dihydrochloride (AAPH) 98%
2,2'-Azobis(2-methylpropionamidine)dihydrochloride (AAPH)
Formula: C₈H₁₈N₆·2HCl
MW: 271.19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00142725
CAS Number: 2997-92-4
UN: 3088
ADR: 4.2,II

Order Now

Specification Test Results

Appearance White to off-white Granules
Infrared spectrum Conforms
Titration Argentometric 97.5 to 102.5 % (on dry substance)
Loss on drying ≤1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR