2-Methyl-3-phenylpropanoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: α-Methylhydrocinnamic acid

37908-0010 37908-0050
TS37908-0010EA 70 USD
TS37908-0010 TS37908-0050
2-Methyl-3-phenylpropanoic acid 98%
2-Methyl-3-phenylpropanoic acid
Formula: C₁₀H₁₂O₂
MW: 164.204 g/mol
Boiling Pt: 167…168 °C (23 mmHg)
Melting Pt: 37…41 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00192301
CAS Number: 1009-67-2

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystals or crystalline mass and/or chunks
Infrared spectrum Conforms
Melting point 37 to 41 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR