Benzyl(triphenylphosphoranylidene)acetate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Benzyl 2-(triphenylphosphoranylidene)acetate
37722-0050
37722-0250
37722-1000
TS37722-0050EA
69
USD
TS37722-0050
TS37722-0250
TS37722-1000
Benzyl(triphenylphosphoranylidene)acetate 97%
Benzyl(triphenylphosphoranylidene)acetate
Formula:
C₂₇H₂₃O₂P MW: 410.453 g/mol Melting Pt: 120…127 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00191787 CAS Number: 15097-38-8 UN: 3464 ADR: 6.1,III |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 120 to 127 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...