(R)-2-Isopropylsuccinic acid-1-methyl ester ≥95%, ee 98%
Supplier: Thermo Scientific Chemicals
36306-0050
36306-5000
TS36306-0050EA
799
USD
TS36306-0050
TS36306-5000
(R)-2-Isopropylsuccinic acid-1-methyl ester ≥95%, ee 98%
(R)-2-Isopropylsuccinic acid-1-methyl ester
Formula:
C₈H₁₄O₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD01091030 CAS Number: 220498-08-8 |
Specification Test Results
Appearance | Clear yellow Viscous liquid |
Infrared spectrum | Conforms |
GC | ≥94 % |
Enantiomeric excess | ≥97.5 % (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...