4-Formylcinnamic acid predominantly trans 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Formylcinnamic acid, predominantly trans, 3-(4-Formylphenyl)acrylic acid

17659-0010 17659-0050 17659-0250
TS17659-0010EA 44 USD
TS17659-0010 TS17659-0050 TS17659-0250
4-Formylcinnamic acid predominantly trans 98%
4-Formylcinnamic acid
Formula: C₁₀H₈O₃
MW: 176.171 g/mol
Melting Pt: 250…255 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006955
CAS Number: 23359-08-2

Order Now

Specification Test Results

Appearance Light yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 250 to 255 °C
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR