2-Methylpropane-1-sulphonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Isobutanesulphonyl chloride, 1-Propanesulphonylchloride, 2-methyl-
43841-0010
43841-0050
TS43841-0010EA
81
USD
TS43841-0010
TS43841-0050
2-Methylpropane-1-sulphonyl chloride 97%
2-Methylpropane-1-sulfonyl chloride
Formula:
C₄H₉ClO₂S Boiling Pt: 77…78 °C (11Torr) Flash Pt: 88 °C Storage Temperature: Ambient |
MDL Number:
MFCD07369800 CAS Number: 35432-36-1 UN: 3265 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to brown Liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...