1-Boc-3-(bromomethyl)azetidine 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: tert-Butyl 3-(bromomethyl)azetidine-1-carboxylate

45216-0010 45216-0050
TS45216-0010EA 243 USD
TS45216-0010 TS45216-0050
1-Boc-3-(bromomethyl)azetidine 95%
1-Boc-3-(bromomethyl)azetidine
Formula: C₉H₁₆BrNO₂
MW: 250.136 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16556174
CAS Number: 253176-93-1

Order Now

Specification Test Results

Appearance White to yellow Low melting solid
Infrared spectrum Conforms
GC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR