3,3,3-trifluoro-2-(trifluoromethyl)propanoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,3,3-Trifluoro-2-(trifluoromethyl)propionic acid

39786-0010 39786-0050
TS39786-0010EA 34 USD
TS39786-0010 TS39786-0050
3,3,3-trifluoro-2-(trifluoromethyl)propanoic acid 97%
3,3,3-trifluoro-2-(trifluoromethyl)propanoic acid
Formula: C₄H₂F₆O₂
MW: 196.048 g/mol
Melting Pt: 50…53 °C
Storage Temperature: Ambient
MDL Number: MFCD00464165
CAS Number: 564-10-3

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 49 to 53 °C
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR