2-Chloro-5-(trifluoromethyl)pyrazine 97%

Supplier: Thermo Scientific Chemicals
Danger

44648-0010 44648-0050
TS44648-0010EA 169 USD
TS44648-0010 TS44648-0050
2-Chloro-5-(trifluoromethyl)pyrazine 97%
2-Chloro-5-(trifluoromethyl)pyrazine
Formula: C₅H₂ClF₃N₂
Boiling Pt: 88 °C (118 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD13189478
CAS Number: 799557-87-2
UN: 2922
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow to orange to brown Liquid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR