N-Boc-4-methylene-L-proline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(S)-1-(tert-Butoxycarbonyl)-4-methylenepyrrolidine-2-carboxylic acid
44926-0010
44926-0050
TS44926-0010EA
88
USD
TS44926-0010
TS44926-0050
N-Boc-4-methylene-L-proline 97%
N-Boc-4-methylene-L-proline
Formula:
C₁₁H₁₇NO₄ Melting Pt: 110…115 °C Storage Temperature: Ambient |
MDL Number:
MFCD01861787 CAS Number: 84348-38-9 |
Specification Test Results
Appearance | White to beige Powder or solid |
Infrared spectrum | Conforms |
Melting point | 110 to 115 °C |
HPLC | ≥96.0 % |
Specific optical rotation | -53° to -63° (20 °C, 589 nm) (c=0.5, CHCl3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...