6-Chloro-7-iodo-7-deazapurine 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine, 6-Chloro-7-iodo-deazapurine

44885-0010 44885-0050
TS44885-0010EA 118 USD
TS44885-0010 TS44885-0050
6-Chloro-7-iodo-7-deazapurine 96%
6-Chloro-7-iodo-7-deazapurine
Formula: C₆H₃ClIN₃
MW: 279.47 g/mol
Melting Pt: 179…183 °C
Storage Temperature: Ambient
MDL Number: MFCD09263258
CAS Number: 123148-78-7

Order Now

Specification Test Results

Appearance Beige to brown Crystalline powder
Infrared spectrum Conforms
Melting point 173 to 180 °C
HPLC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR