Borane diphenylphosphine complex 94%

Supplier: Thermo Scientific Chemicals
Danger

38075-0010 38075-0050
TS38075-0010EA 51 USD
TS38075-0010 TS38075-0050
Borane diphenylphosphine complex 94%
Borane diphenylphosphine complex
Formula: C₁₂H₁₁BP
MW: 197.004 g/mol
Boiling Pt: 148…150 °C (10 mmHg)
Melting Pt: 47…50 °C
Storage Temperature: Ambient
CAS Number: 41593-58-2
UN: 2813
ADR: 4.3,II

Order Now

Specification Test Results

Appearance White to off-white Powder or crystals
Infrared spectrum Conforms
Elemental Analysis ≥92.0 % (ex C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR