O-(6-Chlorobenzotriazol-1-yl)-N,N,N',N'-tetramethyluronium Hexafluorophosphate 98%

Supplier: Thermo Scientific Chemicals
Warning

44893-0100 44893-0500
TS44893-0100EA 51 USD
TS44893-0100 TS44893-0500
O-(6-Chlorobenzotriazol-1-yl)-N,N,N',N'-tetramethyluronium Hexafluorophosphate 98%
O-(6-Chlorobenzotriazol-1-yl)-N,N,N',N'-tetramethyluronium Hexafluorophosphate
Formula: C₁₁H₁₅ClF₆N₅OP
Melting Pt: 185…190 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12031645
CAS Number: 330645-87-9

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 185 to 191 °C
Loss on drying ≤0.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR