N-Methoxy-N-methylcarbamoyl chloride 95%

Supplier: Thermo Scientific Chemicals
Danger

37887-0050 37887-0250
TS37887-0050EA 233 USD
TS37887-0050 TS37887-0250
N-Methoxy-N-methylcarbamoyl chloride 95%
N-Methoxy-N-methylcarbamoyl chloride
Formula: C₃H₆ClNO₂
MW: 123.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06658962
CAS Number: 30289-28-2
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4420 to 1.4440 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR