Naphthenic acid pract.

Supplier: Thermo Scientific Chemicals
Warning

41528-0025 41528-1000 41528-5000
TS41528-0025EA 365 USD
TS41528-0025 TS41528-1000 TS41528-5000
Naphthenic acid pract.
Naphthenic acid
Boiling Pt: 140…200 °C (760 mmHg)
Density: 0.91 g/cm³
Flash Pt: 149 °C
Storage Temperature: Ambient
MDL Number: MFCD00147696
CAS Number: 1338-24-5

Order Now

Specification Test Results

Appearance Clear colorless to yellow to brown Liquid
Infrared spectrum Conforms
Density (20°C) 910 to 940 g/l
Viscosity ≤5 St (at 25°C)
Acid value 230 to 260 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR