Methyl-4-(chloroformyl)benzoate 90% tech.

Supplier: Thermo Scientific Chemicals
Danger

42471-0010 42471-0050
TS42471-0010EA 60 USD
TS42471-0010 TS42471-0050
Methyl-4-(chloroformyl)benzoate 90% tech.
Methyl-4-(chloroformyl)benzoate
Formula: C₉H₇ClO₃
Melting Pt: 50…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00059486
CAS Number: 7377-26-6
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance Off-white to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 50 to 59 °C
Titration RCOCl ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR