Isonicotinoyl chloride hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Pyridine-4-carbonyl Chloride Hydrochloride, 4-Pyridinoyl chloride hydrochloride

21168-0250 21168-1000 21168-2500
TS21168-0250EA 80 USD
TS21168-0250 TS21168-1000 TS21168-2500
Isonicotinoyl chloride hydrochloride 97%
Isonicotinoyl chloride hydrochloride
Formula: C₆H₄ClNO
MW: 178.01 g/mol
Melting Pt: 153…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00012830
CAS Number: 39178-35-3
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 153 to 161 °C
Titration Argentometric ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR