Guanylurea sulphate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dicyandiamidine Sulphate

41114-0250 41114-1000 41114-5000
TS41114-0250EA 102 USD
TS41114-0250 TS41114-1000 TS41114-5000
Guanylurea sulphate 98%
Guanylurea sulfate
Formula: C₂H₈N₄O₅S
Melting Pt: 190 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00035565
CAS Number: 591-01-5

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Water ≤8 % (K.F.)
Titration with NaOH ≥97.5 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR