Di-tert-butyl-N,N-diisopropylphosphoramidite 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Di-tert-butoxy(diisopropylamino)phosphine, Di-tert-butyl N,N-diisopropylphosphoramidite, Di-tert-butyl diisopropylphosphoramidite

42689-0010 42689-0050
TS42689-0010EA 38 USD
TS42689-0010 TS42689-0050
Di-tert-butyl-N,N-diisopropylphosphoramidite 95%
Di-tert-butyl-N,N-diisopropylphosphoramidite
Formula: C₁₄H₃₂NO₂P
MW: 277.39 g/mol
Boiling Pt: 85…90 °C (.2 mmHg)
Density: 0.879 g/cm³
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00153506
CAS Number: 137348-86-8

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4390 to 1.4410 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR